Learn More
FZD4, Mouse, Clone: 3G7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FZD4.
Brand: Abnova H00008322-M02A.200uL
Description
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. [provided by RefSeq
Sequence: TEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYSpecifications
| FZD4 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_012193 | |
| FZD4 | |
| FZD4 (NP_036325,107 a.a. ∽ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8322 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3G7 | |
| Mouse monoclonal antibody raised against a partial recombinant FZD4. | |
| FZD4 | |
| CD344/EVR1/FEVR/FZD4S/Fz-4/FzE4/GPCR/MGC34390 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.