missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH, Mouse, Clone: 1D4, Abnova™
Mouse monoclonal antibody raised against GAPDH.
Brand: Abnova MAB2319.100uL
This item is not returnable.
View return policy
Specifications
| GAPDH | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| GAPDH | |
| GAPDH | |
| Pig GAPDH. | |
| RUO | |
| 396823 | |
| Avian, Bovine, Human, Mouse, Pig, Rat | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1D4 | |
| Mouse monoclonal antibody raised against GAPDH. | |
| In tissue culture supernatant (10mM sodium azide) | |
| GAPD | |
| Mouse | |
| 250 μL | |
| Primary | |
| On Western Blot reveals a prominent ∼37 KDa band. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction