Learn More
GFAP, Mouse, Clone: GA5, Abnova™
Mouse monoclonal antibody raised against GFAP.
Brand: Abnova MAB11287.100ug
Description
This gene encodes one of the major intermediate filament proteins of mature astrocytes. It is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
| GFAP | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) Western Blot (1-3 ug/ml) The optimal working dilution should be determined by the end user. | |
| GFAP | |
| GFAP | |
| Glial fibrillary protein from porcine spinal cord | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| GA5 | |
| Mouse monoclonal antibody raised against GFAP. | |
| In 0.2mg/mL PBS (0.05% BSA, 0.05% sodium azide) | |
| FLJ45472 | |
| Mouse | |
| Protein G affinity chromatography | |
| RUO | |
| 2670 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.