Learn More
GFAP, Mouse, HRP, Clone: SB61b, Abnova™
Mouse monoclonal antibody raised against recombinant GFAP.
Brand: Abnova MAB5897.1mL
Description
This gene encodes one of the major intermediate filament proteins of mature astrocytes. It is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
| GFAP | |
| Monoclonal | |
| Lot specific | |
| Mouse monoclonal antibody raised against recombinant GFAP. | |
| In PBS, pH 7.4 (50% glycerol) | |
| FLJ45472 | |
| Mouse | |
| 1 mL | |
| Primary | |
| human GFAP (∼ 50 KDa). | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| SB61b | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| GFAP | |
| GFAP | |
| Recombinant protein corresponding to human GFAP. | |
| RUO | |
| 2670 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.