missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLYATL2, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GLYATL2.
Brand: Abnova H00219970-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MLVLHNSQKLQILYKSLEKSIPESIKVYGAIFNIKDKNPFNMEVLVDAWPDYQIVITRPQKQEMKDDQDHYTNTYHIFTKASDKLEEVLSYSNVISWEQTLQIQGCQEGLDEAIRKVATSKSVQVDYMKTILFIPELPKKHKTSSNDKMELFEVDDDNKKGNFSNMFIDASHAGLVNEHWAFGKNERSLKYIERCLQDFLGFGVLGPEGQLVSWIVMEQSCELRMGYTVPKYRHQGNMLQIGYHLEKYLSQKEIPFYFHVADNNEKSLQALNNLGFKICPCGWHQWKCTPKKYCSpecifications
| GLYATL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC016789.1 | |
| GLYATL2 | |
| GLYATL2 (AAH16789.1,1 a.a. ∽ 294 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1C5 | |
| Mouse monoclonal antibody raised against a full-length recombinant GLYATL2. | |
| GLYATL2 | |
| BXMAS2-10/GATF-B/MGC24009 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 219970 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction