Learn More
GOLM1, Mouse, Clone: 5B10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GOLM1.
Brand: Abnova H00051280-M06A.200uL
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes proteins synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this gene has been observed to be upregulated in response to viral infection. Alternatively spliced transcript variants encoding the same protein have been described for this gene. [provided by RefSeq
Sequence: VQAALSVSQENPEMEGPERDQLVIPDGQEEEQEAAGEGRNQQKLRGEDDYNMDENEAESETDKQAALAGNDRNIDVFNVEDQKRDTINLLDQREKRNHTLSpecifications
| GOLM1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_016548 | |
| GOLM1 | |
| GOLM1 (NP_057632,302 a.a. ∽ 401 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51280 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5B10 | |
| Mouse monoclonal antibody raised against a partial recombinant GOLM1. | |
| GOLM1 | |
| C9orf155/FLJ22634/FLJ23608/GOLPH2/GP73/PSEC0257/bA379P1.3 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.