missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulocyte, Mouse, Clone: BM-2, Abnova™
Mouse monoclonal antibody raised against Granulocyte.
Brand: Abnova MAB11285.100ug
This item is not returnable.
View return policy
Specifications
| Granulocyte | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein G affinity chromatography | |
| RUO | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| BM-2 | |
| Mouse monoclonal antibody raised against Granulocyte. | |
| In 0.2mg/mL PBS (0.05% BSA, 0.05% sodium azide) | |
| Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction