Learn More
GRB2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant GRB2.
Brand: Abnova H00002885-A01.50uL
Description
The protein encoded by this gene binds the epidermal growth factor receptor and contains one SH2 domain and two SH3 domains. Its two SH3 domains direct complex formation with proline-rich regions of other proteins, and its SH2 domain binds tyrosine phosphorylated sequences. This gene is similar to the Sem5 gene of C. elegans, which is involved in the signal transduction pathway. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MEAIAKYDFKATADDELSFKRGDILKVLNEECDQNWYKAELNGKDGFIPKNYIEMKPHPWFFGKIPRAKAEEMLSKQRHDGAFLIRESESAPGDFSLSVKFGNDVQHFKVLRDGAGKYFLWVVKFNSLNELVDYHRSTSVSRNQQIFLRDIEQVPQQPTYVQALFDFDPQEDGELGFRRGDFIHVMDNSDPNWWKGACHGQTGMFPRNYVTPVNRNVSpecifications
| GRB2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant GRB2. | |
| GRB2 | |
| ASH/EGFRBP-GRB2/Grb3-3/MST084/MSTP084 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| BC000631 | |
| GRB2 | |
| GRB2 (AAH00631, 1 a.a. to 217 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 2885 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only