Learn More
GSK3A Mouse anti-Human, Clone: 6G2F1, Abnova™
Mouse monoclonal antibody raised against partial recombinant GSK3A.
Brand: Abnova MAB10671.100uL
Description
Glycogen synthase kinase 3-alpha (GSK3A; EC {2.7.1.37}) is a multifunctional protein serine kinase, homologous to Drosophila 'shaggy' (zeste-white3) and implicated in the control of several regulatory proteins including glycogen synthase (see GYS1, {138570}) and transcription factors (e.g., JUN, {165160}). It also plays a role in the WNT ({164820}) and PI3K (see PIK3CG, {601232}) signaling pathways (see review by {1:Ali et al. (2001)}).[supplied by OMIM
Specifications
| GSK3A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| GSK3A | |
| GSK3A | |
| Recombinant protein corresponding to human GSK3A. | |
| RUO | |
| 2931 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| 6G2F1, 6G2F2 | |
| Mouse monoclonal antibody raised against partial recombinant GSK3A. | |
| In ascites (0.03% sodium azide) | |
| DKFZp686D0638 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.