missing translation for 'onlineSavingsMsg'
Learn More

H-2Kd/H-2Dd, Mouse, Clone: 34-1-2S, Abnova™

Product Code. 16210820
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16210820 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16210820 Supplier Abnova Supplier No. MAB3259.500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against H-2Kd/H-2Dd.

Specifications

Antigen H-2Kd/H-2Dd
Applications Flow Cytometry
Classification Monoclonal
Clone 34-1-2S
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against H-2Kd/H-2Dd.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Mouse
Immunogen C3H alphaBDF1 splenocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity An epitope in the alpha3 domain that is common to H-2Kd and H-2Dd.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.