missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2-D2, Mouse, PE, Clone: 27-11-13S, Abnova™
Mouse monoclonal antibody raised against H2-D2.
Brand: Abnova MAB5825.100ug
This item is not returnable.
View return policy
Specifications
| H2-D2 | |
| Monoclonal | |
| PE | |
| Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| 27-11-13S | |
| Mouse monoclonal antibody raised against H2-D2. | |
| In PBS (0.09% sodium azide) | |
| C3H alphaBDF1 splenocytes. | |
| RUO | |
| An epitope on the alpha chain of H-2Db. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction