missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA (A/Anhui/1/2005)(H5N1), Mouse, Clone: IT-1031, Abnova™
Mouse monoclonal antibody raised against partial recombinant HA (H5N1).
Brand: Abnova MAB10131.100ug
This item is not returnable.
View return policy
Specifications
| HA (A/Anhui/1/2005)(H5N1) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Affinity purification | |
| RUO | |
| No cross-reactivity to other H5N1 subtypes. | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| IT-1031 | |
| Mouse monoclonal antibody raised against partial recombinant HA (H5N1). | |
| In PBS | |
| Recombinant protein corresponding to amino acids 18-530 of HA (A/Anhui/1/2005)(H5N1). | |
| 100 μg | |
| Primary | |
| Virus | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction