missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Anti-Hamster IgG Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB011
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Hamster samples. It is validated for Western Blot.
Specifications
| IgG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1-2 ug/mL | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3500 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| MAH1.12 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| IGHG1 | |
| Armenian hamster IgG | |
| 500 μg | |
| Secondary | |
| Detects IgG from Armenian Hamster. | |
| Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction