missing translation for 'onlineSavingsMsg'
Learn More

HARS (M03A), Mouse anti-Human, Clone: 4D4, Abnova™

Product Code. 16149174
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16149174 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16149174 Supplier Abnova Supplier No. H00003035M03A.200uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant HARS.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis. [provided by RefSeq

Sequence: MAERAALEELVKLQGERVRGLKQQKASAELIEEEVAKLLKLKAQLGPDESKQKFVLKTPKGTRDYSPRQMAVREKVFDVIIRCFKRHGAEVIDTPV

Specifications

Antigen HARS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HARS.
Formulation ascites with no preservative
Gene HARS
Gene Accession No. NM_002109
Gene Alias FLJ20491/HRS
Gene Symbols HARS
Host Species Mouse
Immunogen HARS (NP_002100,1 a.a. ∽ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3035
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.