missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBV surface Ag Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native HBV surface Ag.
Brand: Abnova MAB1421.1mg
This item is not returnable.
View return policy
Specifications
| HBV surface Ag | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native HBV surface Ag. | |
| In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) | |
| Native purified HBV surface Ag. | |
| RUO | |
| This antibody reacts with the hepatitis B virus e antigen. | |
| Liquid |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction