Learn More
HBXIP, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant HBXIP.
Brand: Abnova H00010542-A01.50uL
Description
This gene encodes a protein that specifically complexes with the C-terminus of hepatitis B virus X protein (HBx). The function of this protein is to negatively regulate HBx activity and thus to alter the replication life cycle of the virus. [provided by RefSeq
Sequence: MEATLEQHLEDTMKNPSIVGVLCTDSQGLNLGCRGTLSDEHAGVISVLAQQAAKLTSDPTDIPVVCLESDNGNIMIQKHDGITVAAHKMASSpecifications
| HBXIP | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant HBXIP. | |
| HBXIP | |
| MGC71071/XIP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC062619 | |
| HBXIP | |
| HBXIP (AAH62619, 83 a.a. ∽ 173 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 10542 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only