missing translation for 'onlineSavingsMsg'
Learn More

Hepatitis B core antigen, Mouse, Clone: 13A9, Abnova™

Product Code. 16056456
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16056456 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16056456 Supplier Abnova Supplier No. MAB4774.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant hepatitis B core antigen.

Specifications

Antigen Hepatitis B core antigen
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 13A9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant hepatitis B core antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Recombinant protein corresponding to hepatitis B core antigen.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HBcAg core antigen. Amino acids 135-140 core antigen. Reacts with synthetic HBc peptides.
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.