missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C virus NS-3, Mouse, Clone: 1-5, Abnova™
Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS-3.
Brand: Abnova MAB6772.500ug
This item is not returnable.
View return policy
Specifications
| Hepatitis C virus NS-3 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (4 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA, Western Blot | |
| 1-5 | |
| Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS-3. | |
| Lyophilized from 10mM PBS, pH 7.2 | |
| Recombinant protein corresponding to least 60 residues in length of Chinese HCV NS-3. | |
| RUO | |
| This antibody has no cross reaction with HCV capsid region and other non-structural regions can be seen. | |
| Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction