missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C virus NS-3, Mouse, Clone: BDI371, Abnova™
Mouse monoclonal antibody raised against recombinant hepatitis C virus NS-3.
Brand: Abnova MAB4794.1mg
This item is not returnable.
View return policy
Specifications
| Hepatitis C virus NS-3 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| BDI371 | |
| Mouse monoclonal antibody raised against recombinant hepatitis C virus NS-3. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Recombinant protein corresponding to HCV NS-3 (genotype 1a). | |
| RUO | |
| Recognizes Hepatitis C virus. Specific to NS-3. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction