missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpesvirus 8 ORF73, Mouse, Clone: AT4C11, Abnova™
Mouse monoclonal antibody raised against partial recombinant human herpesvirus 8 ORF73.
Brand: Abnova MAB4201.100uL
This item is not returnable.
View return policy
Specifications
| Herpesvirus 8 ORF73 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| HHV8GK18_gp81 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| AT4C11 | |
| Mouse monoclonal antibody raised against partial recombinant human herpesvirus 8 ORF73. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| HHV8GK18_gp81 | |
| Recombinant protein corresponding to amino acids 122-329 of human herpesvirus type 8 ORF73. | |
| RUO | |
| 4961527 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction