Learn More
HK2, Mouse, Clone: 3D3, Abnova™
Mouse monoclonal antibody raised against partial recombinant HK2.
Brand: Abnova MAB10684.100uL
Description
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes hexokinase 2, the predominant form found in skeletal muscle. It localizes to the outer membrane of mitochondria. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells. [provided by RefSeq
Specifications
| HK2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| HK2 | |
| HK2 | |
| Recombinant protein corresponding to human HK2. | |
| RUO | |
| 3099 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
| 3D3 | |
| Mouse monoclonal antibody raised against partial recombinant HK2. | |
| In ascites (0.03% sodium azide) | |
| DKFZp686M1669/HKII/HXK2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.