missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR, Mouse, PerCP, Clone: MEM-12, Abnova™
Mouse monoclonal antibody raised against HLA-DR.
Brand: Abnova MAB4482.100Reactions
This item is not returnable.
View return policy
Specifications
| HLA-DR | |
| Monoclonal | |
| PerCP | |
| Flow Cytometry (10 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| HLA-DRA1 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody recognizes common epitope on human HLA-DR which is dependent on the association of alpha and beta chains. DR is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| MEM-12 | |
| Mouse monoclonal antibody raised against HLA-DR. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| HLA-DRA | |
| Native from thymocyte membrane. | |
| RUO | |
| 3122/3123/3125/3126/3127 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction