Learn More
HNT, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HNT.
Brand: Abnova H00050863-A01.50uL
Description
This gene encodes a member of the IgLON (LAMP, OBCAM, Ntm) family of immunoglobulin (Ig) domain-containing glycosylphosphatidylinositol (GPI)-anchored cell adhesion molecules. The encoded protein may promote neurite outgrowth and adhesion via a homophilic mechanism. This gene is closely linked to a related family member, opioid binding protein/cell adhesion molecule-like (OPCML), on chromosome 11. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: VPVRSGDATFPKAMDNVTVRQGESATLRCTIDNRVTRVAWLNRSTILYAGNDKWCLDPRVVLLSNTQTQYSIEIQNVDVYDEGPYTCSVQTDNHPKTSRVHLIVQVSPKISpecifications
| HNT | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant HNT. | |
| HNT | |
| MGC60329/NTM | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016522 | |
| HNT | |
| HNT (NP_057606, 29 a.a. ∽ 138 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 50863 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only