missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA4/HSPA8, Mouse, Clone: N27F3-4, Abnova™
Mouse monoclonal antibody raised against recombinant HSPA4/HSPA8.
Brand: Abnova MAB6130.100ug
This item is not returnable.
View return policy
Specifications
| HSPA4/HSPA8 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (1-10 ug/mL) Immunoprecipitation (1-10 ug/mL) Flow cytometry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| APG-2/HS24/P52/MGC131852/RY/hsp70/hsp70RY | |
| Mouse | |
| 100 μg | |
| Primary | |
| The epitope is in the region of amino acids 503-640 of human Hsp70. | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| N27F3-4 | |
| Mouse monoclonal antibody raised against recombinant HSPA4/HSPA8. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| HSPA4 | |
| Recombinant protein corresponding to human HSPA4/HSPA8. | |
| RUO | |
| 3308/3312 | |
| Bovine, Canine, Chicken, Drosophila, Fish, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Nematode, Plant, Pig, Rabbit, Rat, Sheep | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction