Learn More
CD56 Mouse anti-Human, BUV395, Clone: B159, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 740299
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Specifications
| CD56 | |
| Monoclonal | |
| 0.2mg/mL | |
| NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| B159 | |
| Brilliant Ultraviolet 395 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human NK Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only