missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia species LPS, Mouse, Clone: B352M, Abnova™
Mouse monoclonal antibody raised against Chlamydia species LPS.
Brand: Abnova MAB3948.100ug
This item is not returnable.
View return policy
Specifications
| Chlamydia species LPS | |
| Monoclonal | |
| Unconjugated | |
| Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Bacteria | |
| Liquid |
| ELISA | |
| B352M | |
| Mouse monoclonal antibody raised against Chlamydia species LPS. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Native from Chlamydia trachomatis elementary bodies. | |
| RUO | |
| Specific to LPS of Chlamydia. Genus specific. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction