missing translation for 'onlineSavingsMsg'
Learn More

Chlamydia trachomatis MOMP, Mouse, Clone: CL12-717.16, Abnova™

Product Code. 16066896
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16066896 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16066896 Supplier Abnova Supplier No. MAB6158.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.

Specifications

Antigen Chlamydia trachomatis MOMP
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone CL12-717.16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Chlamydia trachomatis elementary bodies.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies recognize Chlamydia trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, L3.
Target Species Bacteria
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.