missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium botulinum Toxin A SNAP-25, Mouse, Clone: B372M, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A SNAP-25.
Brand: Abnova MAB4733.1mL
This item is not returnable.
View return policy
Specifications
| Clostridium botulinum Toxin A SNAP-25 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mL | |
| Primary | |
| Bacteria | |
| Liquid |
| ELISA, Immunohistochemistry | |
| B372M | |
| Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A SNAP-25. | |
| In cell culture supernatant | |
| A synthetic peptide corresponding to amino acids 183-197 of Clostridium botulinum Toxin A SNAP-25. | |
| RUO | |
| Specific for both the intact whole form of SNAP-25 and the C. botulinum Toxin A cleaved form of SNAP-25. Has been found to bind to cleaved and uncleaved SNAP-25 amino acids 183-197 by ELISA and cell staining techniques. Cross-reactivity: SNAP-25 (cleaved by C. botulinum Toxin A) 100% SNAP-25 (uncleaved by C. botulinum Toxin A) 100% VAMP2 0% Syntaxin 1 0% | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction