missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue virus E-D2, Mouse, Clone: 6j778, Abnova™
Mouse monoclonal antibody raised against partial recombinant Dengue virus E-D2.
Brand: Abnova MAB8905.100uL
This item is not returnable.
View return policy
Specifications
| Dengue virus E-D2 | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against partial recombinant Dengue virus E-D2. | |
| In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
| Recombinant protein corresponding to amino acids 52-280 of Dengue virus E-D2. | |
| 100 μL | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Western Blot | |
| 6j778 | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Affinity purification | |
| RUO | |
| This antibody recognizes recombinant protein Dengue virus E-D2. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction