missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entamoeba histolytica HK-9 Mouse anti-, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against Entamoeba histolytica HK-9.
Brand: Abnova MAB6148.100ug
This item is not returnable.
View return policy
Specifications
| Entamoeba histolytica HK-9 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against Entamoeba histolytica HK-9. | |
| In PBS, pH 7.4 | |
| Entamoeba histolytica HK-9. | |
| RUO | |
| These antibodies recognize Entamoeba histolytica trophozoites and cysts. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Bacteria, Protozoa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction