missing translation for 'onlineSavingsMsg'
Learn More

Staphylococcus aureus, Mouse, Clone: 703, Abnova™

Product Code. 16036226
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16036226 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16036226 Supplier Abnova Supplier No. MAB4338.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against Staphylococcus aureus.

Specifications

Antigen Staphylococcus aureus
Applications ELISA
Classification Monoclonal
Clone 703
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Staphylococcus aureus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native Staphylococcus aureus.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specifically recognize peptidoglycan of Staphylococcus aureus, Protein A-negative Staphylococcus aureus and Staphylococcus epidermidis. Does not cross-react with Streptococcus species, Mycoplasma species, or Corynebacteria species.
Target Species Staphylococcus species
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.