missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM1, Mouse, Clone: YUK11, Abnova™
Description
This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq
Specifications
Specifications
| Antigen | ICAM1 |
| Applications | Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | YUK11 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against native ICAM1. |
| Dilution | Flow Cyt (1-5 ug/mL) Immunoprecipitation (5-10 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS (0.1% proclin, 2.0% Block Ace) |
| Gene | ICAM1 |
| Gene Alias | BB2/CD54/P3.58 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?