missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNA1, Mouse, Clone: ST29, Abnova™
Mouse monoclonal antibody raised against IFNA1.
Brand: Abnova MAB7027.100ug
This item is not returnable.
View return policy
Description
Leukocyte interferon is produced predominantly by B lymphocytes. Immune interferon (IFN-gamma; MIM 147570) is produced by mitogen- or antigen-stimulated T lymphocytes.[supplied by OMIM
Specifications
| IFNA1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| IFNA1 | |
| IFNA1 | |
| Human IFNA1. | |
| RUO | |
| 3439 | |
| Human | |
| Liquid |
| Functional Assay | |
| ST29 | |
| Mouse monoclonal antibody raised against IFNA1. | |
| In PBS | |
| IFL/IFN/IFN-ALPHA/IFNA13/IFNA@/MGC138207/MGC138505/MGC138507 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Interferon alpha (ST29). | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction