missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE, Mouse, Clone: 4G7, Abnova™
Mouse monoclonal antibody raised against IgE.
Brand: Abnova MAB6451.100ug
This item is not returnable.
View return policy
Specifications
| IgE | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 4G7 | |
| Mouse monoclonal antibody raised against IgE. | |
| In TBS, pH 8.0 (0.09% sodium azide) | |
| Human IgE. | |
| RUO | |
| This antibody reacts with human IgE. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction