missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG, Mouse, Clone: ICO-97, Abnova™
Mouse monoclonal antibody raised against IgG.
Brand: Abnova MAB6980.100ug
This item is not returnable.
View return policy
Specifications
| IgG | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry | |
| ICO-97 | |
| Mouse monoclonal antibody raised against IgG. | |
| In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate) | |
| Human IgG. | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction