missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG, Mouse, HRP, Clone: EM-07, Abnova™
Mouse monoclonal antibody raised against recombinant IgG.
Brand: Abnova MAB4494.100ug
This item is not returnable.
View return policy
Specifications
| IgG | |
| Monoclonal | |
| 0.1 mg/mL | |
| Mouse monoclonal antibody raised against recombinant IgG. | |
| In PBS (1% BSA, 0.01% thimerosal) | |
| Recombinant Fc fusion protein corresponding to human IgG. | |
| RUO | |
| This antibody reacts with Fc part of human IgG heavy chain and with isolated Fc fragments. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunocytochemistry, Western Blot | |
| EM-07 | |
| HRP | |
| ELISA (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur