missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2, Mouse, Clone: 52G1, Abnova™
Mouse monoclonal antibody raised against native IgG2.
Brand: Abnova MAB8454.1mg
This item is not returnable.
View return policy
Specifications
| IgG2 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 52G1 | |
| Mouse monoclonal antibody raised against native IgG2. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Native purified IgG2 from human myeloma patient serum. | |
| RUO | |
| 52G1 specific to g-2 epitope inside Fc fragment of human IgG2, Fc-region specific.No cross-reaction with human IgG1, IgG3, IgG4, IgA, IgM and IgE. | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction