missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGKC, Mouse, Clone: HP6053, Abnova™
mouse monoclonal antibody raised against IGKC.
Brand: Abnova MAB11486.100ug
This item is not returnable.
View return policy
Specifications
| IGKC | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-2 ug/million cells) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein lysate) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| IGKC | |
| IGKC | |
| Purified human IGKC. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| HP6053 | |
| mouse monoclonal antibody raised against IGKC. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 3514 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction