missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A virus nucleoprotein, Mouse, Clone: 1331, Abnova™
Mouse monoclonal antibody raised against influenza A virus nucleoprotein.
Brand: Abnova MAB4825.1mg
This item is not returnable.
View return policy
Specifications
| Influenza A virus nucleoprotein | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunofluorescence | |
| 1331 | |
| Mouse monoclonal antibody raised against influenza A virus nucleoprotein. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Influenza A Strain: A/Texas. | |
| RUO | |
| Recognizes the Nucleoprotein. Reactive with >50 separate isolates from H1 through H14. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?