missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A virus nucleoprotein, Mouse, Clone: BDI375, Abnova™
Mouse monoclonal antibody raised against influenza A virus nucleoprotein.
Brand: Abnova MAB4823.1mg
This item is not returnable.
View return policy
Specifications
| Influenza A virus nucleoprotein | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunofluorescence | |
| BDI375 | |
| Mouse monoclonal antibody raised against influenza A virus nucleoprotein. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Influenza A virus infected cell lysate. | |
| RUO | |
| Recognizes Influenza A virus. specific to Nucleoprotein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction