missing translation for 'onlineSavingsMsg'
Learn More

ITGB2, Mouse, APC, Clone: MEM-48, Abnova™

Product Code. 16076376
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16076376 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16076376 Supplier Abnova Supplier No. MAB4592.100Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native ITGB2.

The product of this gene belongs to the integrin beta chain family of proteins. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. This gene encodes the integrin beta chain beta 2. A given chain may combine with multiple partners resulting in different integrins. For example, beta 2 combines with the alpha L chain to form the integrin LFA-1, and combines with the alpha M chain to form the integrin Mac-1. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Defects in this gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Specifications

Antigen ITGB2
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-48
Conjugate APC
Description Mouse monoclonal antibody raised against native ITGB2.
Dilution Flow Cytometry (10 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene ITGB2
Gene Alias CD18/LAD/LCAMB/LFA-1/MAC-1/MF17/MFI7
Gene Symbols ITGB2
Host Species Mouse
Immunogen Native purified ITGB2 from leukocytes of patient suffering from LGL-type leukaemia.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Test Specificity This antibody recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta 2 subunit; beta 2 integrin). CD18 is a 90-95 KDa type I transmembrane protein expressed on all leukocytes.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.