Learn More
JARID1D, Mouse, Clone: 4C6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant JARID1D.
Brand: Abnova H00008284-M03.100ug
Description
This gene encodes a protein containing zinc finger domains. A short peptide derived from this protein is a minor histocompatibility antigen which can lead to graft rejection of male donor cells in a female recipient. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: AICKDRRWARVAQRLHYPPGKNIGSLLRSHYERIIYPYEMFQSGANHVQCNTHPFDNEVKDKEYKPHSIPLRQSVQPSKFSSYSRRAKRLQPDPEPTEEDIEKHSpecifications
| JARID1D | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004653.4 | |
| JARID1D | |
| JARID1D (NP_004644.2,127 a.a. ∽ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 4C6 | |
| Mouse monoclonal antibody raised against a partial recombinant JARID1D. | |
| JARID1D | |
| HY/HYA/KDM5D/KIAA0234/SMCY | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 8284 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.