missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD1A, Mouse, Clone: 1E12, Abnova™
Mouse monoclonal antibody raised against partial recombinant JMJD1A.
Brand: Abnova MAB10706.100uL
This item is not returnable.
View return policy
Description
This gene encodes a zinc finger protein that contains a jumonji domain and may play a role in hormone-dependent transcriptional activation. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| JMJD1A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| JMJD1A | |
| JMJD1A | |
| Recombinant protein corresponding to human JMJD1A. | |
| RUO | |
| 55818 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 1E12 | |
| Mouse monoclonal antibody raised against partial recombinant JMJD1A. | |
| In ascites (0.03% sodium azide) | |
| DKFZp686A24246/DKFZp686P07111/JHDM2A/JHMD2A/JMJD1/KDM3A/KIAA0742/TSGA | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction