missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaposi's sarcoma-associated herpesvirus LANA2, Mouse, Clone: CM-A807, Abnova™
Mouse monoclonal antibody raised against recombinant Kaposi's sarcoma-associated herpesvirus LANA2.
Brand: Abnova MAB2328.100uL
This item is not returnable.
View return policy
Specifications
| Kaposi's sarcoma-associated herpesvirus LANA2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| CM-A807 | |
| Mouse monoclonal antibody raised against recombinant Kaposi's sarcoma-associated herpesvirus LANA2. | |
| In PBS (0.09% sodium azide) | |
| Recombinant fusion protein corresponding to Kaposi's sarcoma-associated herpesvirusLANA2. | |
| RUO | |
| This antibody is specific to KSHV LANA2 (residues 143-310 epitope recognition). By Western Blot, this antibody recognizes a viral protein in KSHV infected B-cells (∼66-70 KDa). | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction