missing translation for 'onlineSavingsMsg'
Learn More

KCNK12, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16138786
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16138786 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16138786 Supplier Abnova Supplier No. H00056660A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant KCNK12.

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other nonpore-forming proteins for activity. [provided by RefSeq

Sequence: ERIISLLAFIMRACRERQLRRSGLLPATFRRGSALSEADSLAGWKPSVYH

Specifications

Antigen KCNK12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KCNK12.
Formulation 50% glycerol
Gene KCNK12
Gene Accession No. NM_022055
Gene Alias THIK-2/THIK2
Gene Symbols KCNK12
Host Species Mouse
Immunogen KCNK12 (NP_071338, 166 a.a. ∽ 215 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56660
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.