missing translation for 'onlineSavingsMsg'
Learn More

Kcnt1, Mouse, Clone: S3-26, Abnova™

Product Code. 16201700
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16201700 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16201700 Supplier Abnova Supplier No. MAB7886.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant Kcnt1.

Specifications

Antigen Kcnt1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S3-26
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Kcnt1.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcnt1
Gene Alias Slack/rSlo2
Gene Symbols Kcnt1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1168-1237 of rat Kcnt1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60444
Test Specificity This antibody cross-reacted weakly ith human.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.