Learn More
KIF22, Mouse, Clone: 5F3, Abnova™
Mouse monoclonal antibody raised against partial recombinant KIF22.
Brand: Abnova MAB10710.100uL
Description
The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintenance. [provided by RefSeq
Specifications
| KIF22 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| KIF22 | |
| KIF22 | |
| Recombinant protein corresponding to human KIF22. | |
| RUO | |
| 3835 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 5F3 | |
| Mouse monoclonal antibody raised against partial recombinant KIF22. | |
| In ascites (0.03% sodium azide) | |
| KID/KNSL4/OBP/OBP-1/OBP-2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.