Learn More
KIT, Mouse, PE, Clone: 104D2, Abnova™
Mouse monoclonal antibody raised against KIT.
Brand: Abnova MAB9828.100Reactions
Description
This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| KIT | |
| Monoclonal | |
| PE | |
| Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user. | |
| KIT | |
| KIT | |
| MOLM-1 megakaryocytic cells. | |
| 100 Reactions | |
| Primary | |
| The mouse monoclonal antibody 104D2 detects extracellular part of KIT (CD117 / c-Kit) protooncogen. | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 104D2 | |
| Mouse monoclonal antibody raised against KIT. | |
| In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide) | |
| C-Kit/CD117/PBT/SCFR | |
| Mouse | |
| Size-exclusion chromatography purification | |
| RUO | |
| 3815 | |
| Bovine, Human, Non-human Primate | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.