missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF10, Mouse, Clone: 2E11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KLF10.
Brand: Abnova H00007071-M44.100ug
This item is not returnable.
View return policy
Description
Sequence: MEERMEMISERPKESMYSWNKTAEKSDFEAVEALMSMSCSWKSDFKKYVENRPVTPVSDLSEEENLLPGTPDFHTIPAFCLTPPYSPSDFEPSQVSNLMAPAPSTVHFKSSpecifications
| KLF10 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC011538.1 | |
| KLF10 | |
| KLF10 (AAH11538.1,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E11 | |
| Mouse monoclonal antibody raised against a partial recombinant KLF10. | |
| KLF10 | |
| EGRA/TIEG/TIEG1 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 7071 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction