Learn More
KRT19, Mouse, Clone: 7, Abnova™
Mouse monoclonal antibody raised against full-length recombinant KRT19.
Brand: Abnova MAB9972.100ug
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. Unlike its related family members, this smallest known acidic cytokeratin is not paired with a basic cytokeratin in epithelial cells. It is specifically expressed in the periderm, the transiently superficial layer that envelopes the developing epidermis. The type I cytokeratins are clustered in a region of chromosome 17q12-q21. [provided by RefSeq
Specifications
| KRT19 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:1000) The optimal working dilution should be determined by the end user. | |
| KRT19 | |
| KRT19 | |
| Recombinant protein corresponding to full length human KRT19. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry | |
| 7 | |
| Mouse monoclonal antibody raised against full length recombinant KRT19. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| CK19/K19/K1CS/MGC15366 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 3880 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.